siRNA Details
| Gene Symbol: Pnma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100461.3 | NP_001093931.1 | 1 | 1257 | ||
| GenBank mRNA | AK141385.1 | - | 1 | 257 | ||
| AK220432.1 | - | 1 | 1257 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnma5| Gene Symbol: Pnma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100461.3 | NP_001093931.1 | 1 | 1257 | ||
| GenBank mRNA | AK141385.1 | - | 1 | 257 | ||
| AK220432.1 | - | 1 | 1257 | |||