siRNA Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198927.3 | NP_945121.1 | 5 | 607 | ||
| GenBank mRNA | AK132579.1 | - | 5 | 609 | ||
| AY459348.1 | - | 5 | 608 | |||
| BC055490.1 | - | 5 | 589 | |||
| BC092073.1 | - | 4 | 428 | |||
| BC138091.1 | - | 5 | 608 | 8861110 | ||

Smgc
Genomic Map