siRNA Details
| Gene Symbol: Morn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081100.1 | NP_001074569.1 | 5, 6 | 471 | ||
| GenBank mRNA | AK015161.1 | - | 5, 6 | 446 | ||
| AK045316.1 | - | 5, 6 | 446 | |||
| AK148312.1 | - | 5, 6 | 642 | |||
| BC132619.1 | - | 5, 6 | 466 | 40130896 | ||
| BC145744.1 | - | 5, 6 | 466 | 40131047 | ||

Morn1
Genomic Map