siRNA Details
| Gene Symbol: Morn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081100.1 | NP_001074569.1 | 3 | 208 | ||
| GenBank mRNA | AK015161.1 | - | 3 | 183 | ||
| AK045316.1 | - | 3 | 182 | |||
| AK148312.1 | - | 3 | 379 | |||
| AK204658.1 | - | 2 | 68 | |||
| BC132619.1 | - | 3 | 203 | 40130896 | ||
| BC145744.1 | - | 3 | 203 | 40131047 | ||

Morn1
Genomic Map