siRNA Details
| Gene Symbol: Crygc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082573.2 | NP_001076042.1 | 3 | 466 | ||
| NM_007775.2 | NP_031801.1 | 3 | 463 | |||
| GenBank mRNA | BC056454.1 | - | 2 | 442 | 6824225 | |
| CJ066821.1 | - | 3 | 464 | |||
| CK627566.1 | - | 1 | 61 | |||
| EF426301.1 | - | 3 | 452 | |||
| EF426302.1 | - | 3 | 455 | |||
| M64544.1 | - | 2 | 415 | |||

Crygc
Genomic Map