siRNA Details
| Gene Symbol: Krtap16-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183296.1 | NP_899119.1 | 1 | 510 | ||
| GenBank mRNA | AF345293.1 | - | 1 | 450 | ||
| AK017265.1 | - | 1 | 510 | |||
| BC107303.2 | - | 1 | 446 | |||
| BC107304.2 | - | 1 | 446 | |||
| BC119503.1 | - | 1 | 293 | |||
| BC119504.2 | - | 1 | 446 | |||
| BC127231.1 | - | 1 | 446 | |||
| BC127232.1 | - | 1 | 446 | |||

Krtap16-3
Genomic Map