siRNA Details
| Gene Symbol: Mphosph9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081323.2 | NP_001074792.1 | 9 | 1987 | ||
| NM_001277867.1 | NP_001264796.1 | 10 | 2077 | |||
| GenBank mRNA | AK081178.1 | - | 9 | 1836 | ||
| AK084586.1 | - | 9 | 1821 | |||
| BC076624.1 | - | 9 | 1850 | |||
| BC138423.1 | - | 6 | 1389 | 8861443 | ||
| BC145681.1 | - | 6 | 1389 | 40130984 | ||

Mphosph9
Genomic Map