siRNA Details
| Gene Symbol: Il17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145834.3 | NP_665833.3 | 1 | 141 | ||
| GenBank mRNA | AF458061.1 | - | 1 | 141 | ||
| BC145853.1 | - | 1 | 204 | 40131156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17c| Gene Symbol: Il17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145834.3 | NP_665833.3 | 1 | 141 | ||
| GenBank mRNA | AF458061.1 | - | 1 | 141 | ||
| BC145853.1 | - | 1 | 204 | 40131156 | ||