siRNA Details
| Gene Symbol: Defa-rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007844.2 | NP_031870.2 | 2 | 327 | ||
| GenBank mRNA | AK008584.1 | - | 2 | 328 | ||
| BC117042.1 | - | 2 | 302 | 40126361 | ||
| BC117044.1 | - | 2 | 302 | 40126363 | ||
| BC145556.1 | - | 2 | 332 | 9054120 | ||
| BC171966.1 | - | 2 | 332 | 9054252 | ||
| EU760893.1 | - | 1 | 123 | |||
| EU760894.1 | - | 1 | 123 | |||
| M33226.1 | - | 2 | 307 | |||
| Gene Symbol: Gm14851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177482.1 | NP_001170953.1 | 2 | 282 | ||
| GenBank mRNA | AV063918.1 | - | 1 | 117 | ||
| AV064248.1 | - | 1 | 118 | |||
| AY761183.1 | - | 2 | 282 | |||

Defa-rs1
Genomic Map