siRNA Details
| Gene Symbol: Defa-rs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007844.2 | NP_031870.2 | 2 | 375 | ||
| GenBank mRNA | AK008584.1 | - | 2 | 376 | ||
| BC117042.1 | - | 2 | 350 | 40126361 | ||
| BC117044.1 | - | 2 | 350 | 40126363 | ||
| BC145556.1 | - | 2 | 380 | 9054120 | ||
| BC171966.1 | - | 2 | 380 | 9054252 | ||
| EU760893.1 | - | 1 | 171 | |||
| EU760894.1 | - | 1 | 171 | |||
| M33226.1 | - | 2 | 355 | |||
| Gene Symbol: AY761184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007582.3 | NP_001007583.1 | 2 | 375 | ||
| GenBank mRNA | AK008498.1 | - | 2 | 381 | ||
| AY761184.1 | - | 2 | 330 | |||
| EU760895.1 | - | 1 | 171 | |||
| Gene Symbol: Gm14851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177482.1 | NP_001170953.1 | 2 | 330 | ||
| GenBank mRNA | AV063918.1 | - | 1 | 165 | ||
| AV064248.1 | - | 1 | 166 | |||
| AY761183.1 | - | 2 | 330 | |||

Defa-rs1
Genomic Map