siRNA Details
| Gene Symbol: Klra7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110323.1 | NP_001103793.1 | 3, 4 | 338 | ||
| NM_014194.5 | NP_055009.5 | 2, 3 | 278 | |||
| GenBank mRNA | AK143240.1 | - | 2, 3 | 238 | ||
| EU429321.1 | - | 2, 3 | 208 | |||
| U10093.1 | - | 3, 4 | 338 | |||
| U10094.1 | - | 2, 3 | 278 | |||
| U10095.1 | - | 3, 4 | 338 | |||
| U12890.1 | - | 2, 3 | 229 | |||

Klra7
Genomic Map