siRNA Details
| Gene Symbol: Gm14459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126491.1 | NP_001119963.1 | 5 | 364 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14459| Gene Symbol: Gm14459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126491.1 | NP_001119963.1 | 5 | 364 | ||