siRNA Details
| Gene Symbol: Rc3h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100591.1 | NP_001094061.1 | 8 | 1217 | ||
| NM_001290642.1 | NP_001277571.1 | 8 | 1217 | |||
| GenBank mRNA | AK053071.1 | - | 8 | 1217 | ||
| BC117951.1 | - | 8 | 1210 | 40098089 | ||
| BC117952.1 | - | 8 | 1210 | 40098090 | ||

Rc3h2
Genomic Map