siRNA Details
| Gene Symbol: Ccna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007628.3 | NP_031654.2 | 7 | 1173 | ||
| GenBank mRNA | AK077114.1 | - | 7 | 1179 | ||
| BB610316.1 | - | 2 | 164 | |||
| BC120518.1 | - | 6 | 1088 | 40129441 | ||
| BC125436.1 | - | 6 | 1088 | 40129951 | ||
| X84311.1 | - | 7 | 1172 | |||

Ccna1
Genomic Map