siRNA Details
| Gene Symbol: Sclt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081411.1 | NP_001074880.1 | 4 | 711 | ||
| GenBank mRNA | AK011907.1 | - | 4 | 509 | ||
| AK014777.1 | - | 4 | 535 | |||
| AK083894.1 | - | 4 | 694 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sclt1| Gene Symbol: Sclt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081411.1 | NP_001074880.1 | 4 | 711 | ||
| GenBank mRNA | AK011907.1 | - | 4 | 509 | ||
| AK014777.1 | - | 4 | 535 | |||
| AK083894.1 | - | 4 | 694 | |||