siRNA Details
| Gene Symbol: Sclt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081411.1 | NP_001074880.1 | 12 | 1434 | ||
| GenBank mRNA | AK011907.1 | - | 12 | 1232 | ||
| AK016469.1 | - | 12 | 1423 | |||
| AK083894.1 | - | 10 | 1490 | |||
| BC026498.1 | - | 7 | 590 | |||
| BC043136.1 | - | 12 | 1401 | 6812105 | ||
| BC060271.1 | - | 12 | 1401 | 6812105 | ||

Sclt1
Genomic Map