siRNA Details
| Gene Symbol: BC100530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082546.1 | NP_001076015.1 | 3 | 337 | ||
| GenBank mRNA | BC100530.1 | - | 3 | 515 | 30289147 | |
| Gene Symbol: BC117090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY169235.1 | - | 3 | 304 | ||
| BC117064.1 | - | 3 | 304 | 40126383 | ||
| BC117090.1 | - | 3 | 304 | 40126409 | ||

BC100530
Genomic Map