siRNA Details
| Gene Symbol: Focad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081184.1 | NP_001074653.1 | 43 | 5357 | ||
| GenBank mRNA | AK122556.1 | - | 16 | 5551 | ||
| AK146138.1 | - | 14 | 1739 | |||
| AK146256.1 | - | 14 | 1739 | |||
| BC039622.1 | - | 5 | 738 | |||
| BC057079.1 | - | 32 | 3835 | |||
| BC089005.1 | - | 41 | 5023 | |||

Focad
Genomic Map