siRNA Details
| Gene Symbol: Trim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053166.2 | NP_444396.2 | 5 | 577 | ||
| GenBank mRNA | AF396656.1 | - | 5 | 592 | ||
| AK029860.1 | - | 204 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim7| Gene Symbol: Trim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053166.2 | NP_444396.2 | 5 | 577 | ||
| GenBank mRNA | AF396656.1 | - | 5 | 592 | ||
| AK029860.1 | - | 204 | ||||