siRNA Details
| Gene Symbol: Cdk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009874.3 | NP_034004.2 | 10 | 928 | ||
| GenBank mRNA | AK145759.1 | - | 10 | 854 | ||
| AK168286.1 | - | 10 | 927 | |||
| BC004605.1 | - | 9 | 775 | 3585145 | ||
| BC068160.1 | - | 10 | 913 | 30358255 | ||
| BC141043.1 | - | 10 | 885 | 9055655 | ||
| U11822.1 | - | 10 | 891 | |||
| X74145.1 | - | 10 | 910 | |||

Cdk7
Genomic Map