siRNA Details
| Gene Symbol: Zfp831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099328.1 | NP_001092798.1 | 1 | 3557 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp831| Gene Symbol: Zfp831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099328.1 | NP_001092798.1 | 1 | 3557 | ||