siRNA Details
| Gene Symbol: Tusc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026954.1 | NP_081230.1 | 1 | 1216 | ||
| GenBank mRNA | AK008612.1 | - | 1 | 927 | ||
| AY546089.1 | - | 1 | 1216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tusc1| Gene Symbol: Tusc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026954.1 | NP_081230.1 | 1 | 1216 | ||
| GenBank mRNA | AK008612.1 | - | 1 | 927 | ||
| AY546089.1 | - | 1 | 1216 | |||