siRNA Details
Gene Symbol: Tusc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026954.1 | NP_081230.1 | 1 | 1268 | ||
GenBank mRNA | AK008612.1 | - | 1 | 979 | ||
AY546089.1 | - | 1 | 1268 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tusc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026954.1 | NP_081230.1 | 1 | 1268 | ||
GenBank mRNA | AK008612.1 | - | 1 | 979 | ||
AY546089.1 | - | 1 | 1268 |