siRNA Details
| Gene Symbol: Vmn2r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175674.3 | NP_783605.2 | 2 | 1133 | ||
| GenBank mRNA | AK030224.1 | - | 2232 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r7| Gene Symbol: Vmn2r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175674.3 | NP_783605.2 | 2 | 1133 | ||
| GenBank mRNA | AK030224.1 | - | 2232 | |||
Set Membership: |
|