siRNA Details
| Gene Symbol: Gpr137c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027518.2 | NP_081794.2 | 4 | 1114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr137c| Gene Symbol: Gpr137c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027518.2 | NP_081794.2 | 4 | 1114 | ||