siRNA Details
| Gene Symbol: Gm5132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085517.2 | NP_001078986.1 | 1 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5132| Gene Symbol: Gm5132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085517.2 | NP_001078986.1 | 1 | 372 | ||