siRNA Details
| Gene Symbol: Gm5132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085517.2 | NP_001078986.1 | 1 | 347 | ||
| Gene Symbol: Gm5382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034100.1 | NP_001029272.1 | 1 | 365 | ||
| GenBank mRNA | BC100482.1 | - | 1 | 365 | 30918325 | |

Gm5132
Genomic Map