siRNA Details
| Gene Symbol: Kmt2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081383.1 | NP_001074852.1 | 38 | 8985 | ||
| GenBank mRNA | AK033338.1 | - | 1 | 264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kmt2c| Gene Symbol: Kmt2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081383.1 | NP_001074852.1 | 38 | 8985 | ||
| GenBank mRNA | AK033338.1 | - | 1 | 264 | ||