siRNA Details
Gene Symbol: Fancf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115087.1 | NP_001108559.1 | 1 | 1441 | ||
GenBank mRNA | AK139575.1 | - | 1 | 912 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fancf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115087.1 | NP_001108559.1 | 1 | 1441 | ||
GenBank mRNA | AK139575.1 | - | 1 | 912 |