siRNA Details
| Gene Symbol: Mettl7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199477.2 | NP_955771.2 | 2 | 1439 | ||
| GenBank mRNA | AB253944.1 | - | 2 | 1430 | ||
| AK143923.1 | - | 2 | 1440 | |||
| BC090971.1 | - | 2 | 1439 | 30314995 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl7a2| Gene Symbol: Mettl7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199477.2 | NP_955771.2 | 2 | 1439 | ||
| GenBank mRNA | AB253944.1 | - | 2 | 1430 | ||
| AK143923.1 | - | 2 | 1440 | |||
| BC090971.1 | - | 2 | 1439 | 30314995 | ||