siRNA Details
| Gene Symbol: Ankle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172756.2 | NP_766344.2 | 6 | 1138 | ||
| GenBank mRNA | AK049395.1 | - | 4 | 1445 | ||
| AK078830.1 | - | 5 | 1368 | |||
| AK080768.1 | - | 4 | 1451 | |||
| AK083552.1 | - | 4 | 1437 | |||
| AK087444.1 | - | 4 | 1456 | |||
| BC030436.1 | - | 6 | 1077 | |||
| BC094316.1 | - | 6 | 1095 | |||
| BC145659.1 | - | 2 | 361 | |||
| EU184014.1 | - | 6 | 1114 | |||

Ankle1
Genomic Map