siRNA Details
| Gene Symbol: Ankle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172756.2 | NP_766344.2 | 4 | 381 | ||
| GenBank mRNA | AK049395.1 | - | 3 | 449 | ||
| AK078830.1 | - | 4 | 372 | |||
| AK080768.1 | - | 3 | 454 | |||
| AK083552.1 | - | 3 | 441 | |||
| AK087444.1 | - | 3 | 455 | |||
| BC030436.1 | - | 4 | 380 | |||
| BC094316.1 | - | 4 | 338 | |||
| EU184014.1 | - | 4 | 357 | |||

Ankle1
Genomic Map