siRNA Details
| Gene Symbol: ZFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015174.1 | NP_055989.1 | 9 | 1450 | ||
| XM_011527831.2 | XP_011526133.1 | 9 | 1449 | |||
| GenBank mRNA | AB029009.1 | - | 8 | 1328 | ||
| AK295037.1 | - | 8 | 1271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFR2| Gene Symbol: ZFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015174.1 | NP_055989.1 | 9 | 1450 | ||
| XM_011527831.2 | XP_011526133.1 | 9 | 1449 | |||
| GenBank mRNA | AB029009.1 | - | 8 | 1328 | ||
| AK295037.1 | - | 8 | 1271 | |||