siRNA Details
Gene Symbol: Mif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010798.2 | NP_034928.1 | 2 | 223 | ||
GenBank mRNA | AK008939.1 | - | 2 | 217 | ||
AK010691.1 | - | 2 | 194 | |||
AK013328.1 | - | 2 | 217 | |||
AK152211.1 | - | 2 | 204 | |||
BC024895.1 | - | 2 | 203 | 3978685 | ||
BC086928.1 | - | 2 | 193 | 6762214 | ||
BC100605.1 | - | 2 | 193 | 30118109 | ||
L02913.1 | - | 2 | 196 | |||
L07607.1 | - | 2 | 126 | |||
L10613.1 | - | 2 | 222 | |||
Z23048.1 | - | 2 | 143 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
