siRNA Details
| Gene Symbol: NBEAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015175.2 | NP_055990.1 | 40 | 6636 | ||
| XM_006713072.3 | XP_006713135.1 | 39 | 6552 | |||
| XM_011533533.2 | XP_011531835.1 | 40 | 6633 | |||
| XM_017006010.1 | XP_016861499.1 | 40 | 6633 | |||
| XM_017006011.1 | XP_016861500.1 | 40 | 6489 | |||
| XM_017006012.1 | XP_016861501.1 | 39 | 6552 | |||
| XM_017006013.1 | XP_016861502.1 | 40 | 6633 | |||
| XM_017006014.1 | XP_016861503.1 | 39 | 6408 | |||
| XM_017006015.1 | XP_016861504.1 | 38 | 6264 | |||
| XM_017006016.1 | XP_016861505.1 | 35 | 7210 | |||
| XM_017006017.1 | XP_016861506.1 | 17 | 3052 | |||
| GenBank mRNA | AB011112.3 | - | 26 | 4320 | ||
| AK074036.1 | - | 1 | 86 | |||
| AK131104.1 | - | 40 | 6476 | |||
| AK301269.1 | - | 5 | 699 | |||
| AY358455.1 | - | 10 | 1586 | |||
| BC029944.1 | - | 2 | 57 | |||
| BC060874.1 | - | 8 | 1114 | |||

NBEAL2
Genomic Map