siRNA Details
| Gene Symbol: Esco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028039.2 | NP_082315.3 | 3 | 987 | ||
| GenBank mRNA | AK083728.1 | - | 1 | 766 | ||
| AK144685.1 | - | 3 | 943 | |||
| BC071275.1 | - | 3 | 920 | 6331551 | ||
| CF730483.1 | - | 1 | 507 | |||

Esco2
Genomic Map