siRNA Details
| Gene Symbol: Esco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028039.2 | NP_082315.3 | 3 | 338 | ||
| GenBank mRNA | AK050845.1 | - | 3 | 289 | ||
| AK083728.1 | - | 1 | 117 | |||
| AK144685.1 | - | 3 | 294 | |||
| BC071275.1 | - | 3 | 271 | 6331551 | ||
| BY029949.1 | - | 3 | 366 | |||
| CF168369.1 | - | 3 | 321 | |||

Esco2
Genomic Map