siRNA Details
| Gene Symbol: Zfp189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289901.1 | NP_001276830.1 | 3 | 286 | ||
| NM_145547.3 | NP_663522.2 | 2 | 457 | |||
| GenBank mRNA | AK049475.1 | - | 2 | 366 | ||
| AK051265.1 | - | 2 | 416 | |||
| BC021326.1 | - | 2 | 419 | 5065942 | ||
| BC087833.1 | - | 2 | 323 | 5708354 | ||
| BY085925.1 | - | 3 | 290 | |||

Zfp189
Genomic Map