siRNA Details
| Gene Symbol: 1700012L04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029588.3 | NP_083864.1 | 1 | 319 | ||
| GenBank mRNA | AK005922.1 | - | 1 | 321 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700012L04Rik| Gene Symbol: 1700012L04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029588.3 | NP_083864.1 | 1 | 319 | ||
| GenBank mRNA | AK005922.1 | - | 1 | 321 | ||