siRNA Details
| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172835.3 | NP_766423.3 | 2 | 277 | ||
| GenBank mRNA | AK044418.1 | - | 2 | 224 | ||
| AK135575.1 | - | 2 | 226 | |||
| BC028931.1 | - | 2 | 235 | 5037308 | ||
| BY128359.1 | - | 2 | 277 | |||

Peli3
Genomic Map