siRNA Details
| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172835.3 | NP_766423.3 | 5 | 565 | ||
| GenBank mRNA | AK044418.1 | - | 5 | 512 | ||
| AK135575.1 | - | 4 | 412 | |||
| BC028931.1 | - | 5 | 523 | 5037308 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Peli3| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172835.3 | NP_766423.3 | 5 | 565 | ||
| GenBank mRNA | AK044418.1 | - | 5 | 512 | ||
| AK135575.1 | - | 4 | 412 | |||
| BC028931.1 | - | 5 | 523 | 5037308 | ||