siRNA Details
| Gene Symbol: Kdm7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033430.4 | NP_001028602.2 | 19 | 2825 | ||
| GenBank mRNA | AK035529.1 | - | 10 | 1839 | ||
| AK129429.1 | - | 8 | 1167 | |||
| AK136169.1 | - | 7 | 964 | |||
| AK136209.1 | - | 19 | 2664 | |||
| AK136238.1 | - | 19 | 2812 | |||
| BC007161.1 | - | 19 | 2521 | |||
| BC145216.1 | - | 19 | 2711 | |||
| BC145848.1 | - | 19 | 2715 | 40131151 | ||

Kdm7a
Genomic Map