siRNA Details
| Gene Symbol: Kdm7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033430.4 | NP_001028602.2 | 10 | 1416 | ||
| GenBank mRNA | AK042327.1 | - | 10 | 1433 | ||
| AK042834.1 | - | 10 | 1417 | |||
| AK136209.1 | - | 10 | 1255 | |||
| AK136238.1 | - | 10 | 1403 | |||
| BC007161.1 | - | 10 | 1112 | |||
| BC145216.1 | - | 10 | 1302 | |||
| BC145848.1 | - | 10 | 1306 | 40131151 | ||

Kdm7a
Genomic Map