siRNA Details
Gene Symbol: Kdm7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033430.4 | NP_001028602.2 | 10 | 1438 | ||
GenBank mRNA | AK042327.1 | - | 10 | 1455 | ||
AK042834.1 | - | 10 | 1439 | |||
AK136209.1 | - | 10 | 1277 | |||
AK136238.1 | - | 10 | 1425 | |||
BC007161.1 | - | 10 | 1134 | |||
BC145216.1 | - | 10 | 1324 | |||
BC145848.1 | - | 10 | 1328 | 40131151 |
