siRNA Details
| Gene Symbol: Vps13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177151.3 | NP_796125.2 | 55 | 10295 | ||
| GenBank mRNA | AK122302.1 | - | 16 | 2866 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps13b| Gene Symbol: Vps13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177151.3 | NP_796125.2 | 55 | 10295 | ||
| GenBank mRNA | AK122302.1 | - | 16 | 2866 | ||