siRNA Details
| Gene Symbol: Akr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145364.2 | NP_663339.1 | 8 | 983 | ||
| GenBank mRNA | BC018333.1 | - | 8 | 983 | 4162788 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akr1d1| Gene Symbol: Akr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145364.2 | NP_663339.1 | 8 | 983 | ||
| GenBank mRNA | BC018333.1 | - | 8 | 983 | 4162788 | |