siRNA Details
Gene Symbol: Gm4836 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009529.3 | NP_033555.1 | 2 | 222 | ||
GenBank mRNA | BC061169.1 | - | 2 | 222 | 6774630 |
Gene Symbol: Gm10058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109969.2 | NP_001103439.1 | 2 | 191 | ||
GenBank mRNA | BU936093.1 | - | 194 |
Gene Symbol: Gm10096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102678.2 | NP_001096148.1 | 2 | 222 |
Gene Symbol: Gm10147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099919.2 | NP_001093389.1 | 194 |
Gene Symbol: Gm10230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099347.2 | NP_001092817.1 | 2 | 191 |
Gene Symbol: Gm10486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109970.1 | NP_001103440.1 | 2 | 222 | ||
GenBank mRNA | BC087930.1 | - | 191 | 6773503 | ||
BC099540.1 | - | 2 | 162 | 6774523 | ||
X72697.1 | - | 209 |
Gene Symbol: Gm10487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100609.1 | NP_001094079.1 | 2 | 222 |
Gene Symbol: Gm10488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099325.2 | NP_001092795.1 | 2 | 191 | ||
GenBank mRNA | BC099541.1 | - | 2 | 191 | 30916030 |
Gene Symbol: Gm14525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162364.2 | NP_001155836.1 | 2 | 191 | ||
GenBank mRNA | BC048742.1 | - | 2 | 191 |
Gene Symbol: Gm14632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100610.2 | NP_001094080.1 | 2 | 191 |
Gene Symbol: Gm14819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110250.2 | NP_001103720.1 | 191 |
Gene Symbol: Gm2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104946.2 | NP_001098416.1 | 2 | 191 |
Gene Symbol: Gm5169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040669.1 | NP_001035759.1 | 2 | 188 | ||
GenBank mRNA | BC087929.1 | - | 2 | 188 | 6742946 |
Gene Symbol: Gm6121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114754.1 | NP_001108226.1 | 2 | 191 |
