siRNA Details
Gene Symbol: Vmn2r66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033878.3 | NP_001029050.3 | 3 | 625 | ||
GenBank mRNA | AK156873.1 | - | 1 | 546 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
