siRNA Details
| Gene Symbol: Vmn2r122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009488.2 | NP_033514.2 | 2 | 441 | ||
| NM_009492.2 | NP_033518.2 | 2 | 441 | |||
| GenBank mRNA | AF011412.1 | - | 2 | 441 | ||
| AF011422.1 | - | 2 | 415 | |||
More Information
Set Membership: |
|

Vmn2r122
Genomic Map