siRNA Details
| Gene Symbol: 1700047I17Rik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100116.1 | NP_001093586.1 | 519 | |||
| Gene Symbol: Fam177a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028527.1 | NP_082803.1 | 4 | 519 | ||
| GenBank mRNA | AK006713.1 | - | 352 | |||
| AK045530.1 | - | 579 | ||||
| BC048158.1 | - | 519 | 5698044 | |||
| BC049669.1 | - | 578 | 6535412 | |||

1700047I17Rik2
Genomic Map